N-tert-Butyl-ɑ-phenylnitrone

Supplier: ADIPOGEN CORP MS

Synonyms: PBN, N-Benzylidene-tert-butylamine N-Oxide, N-tert-Butyl-α-phenylnitrone

CDX-B0269-G001 CDX-B0269-G005
102987-494EA 78.92 USD
102987-494 102987-496
N-tert-Butyl-ɑ-phenylnitrone
N-tert-Butyl-alpha-phenylnitrone

Cell permeable spin trap commonly used in free radical research for both in vivo and in vitro studies. It protects against oxidative damage caused by various inflammatory events, demonstrating neuroprotective, anti-anging, and antidiabetic effects. PBN has been shown to inhibit LPS-induced NF-kappaB DNA binding activity, inhibit COX-2 catalytic activity, inhibit lipid peroxidation in rat liver microsomes and prevent the induction of inducible nitric oxide synthase (iNOS). Shows anti-cancer activity in several experimental cancer models.

Formula: C₁₁H₁₅NO
MDL Number: MFCD00008799
CAS Number: 3376-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR