Aspergillin PZ ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0162-M001
AG-CN2-0162-M005
102987-426EA
207.69
USD
102987-426
102987-428
Aspergillin PZ ≥95%
Aspergillin PZ
Antibiotic
Source/Host: Isolated from Aspergillus sp.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO, methanol or ethanol.
Inducer of morphological deformation of the conidia of Pyricularia oryzae.
Storage Temperature:
Freezer |
CAS Number:
483305-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...