Lactacystin ≥98%

Supplier: ADIPOGEN CORP MS

AG-CN2-0104-C100 AG-CN2-0104-C200 AG-CN2-0104-C500 AG-CN2-0104-M001
102987-354EA 96.92 USD
102987-354 102987-356 102987-358 102987-360
Lactacystin ≥98%
Lactacystin

Induces neuritogenesis. Potent and selective irreversible proteasome inhibitor. Specifically inhibits 20S proteasome (MCP; multicatalytic complex). Blocks proteasome activity by targeting the catalytic beta-subunit. Apoptosis inducer. Anticancer compound. Upregulates HSP70 and HSP22. Induces differentiation and inhibits cell cycle progression in several tumor cell lines. Inhibits NF-kappaB activation by inhibiting IkappaB degradation. Cathepsin A inhibitor. beta-Secretase activity inhibitor. Anti-adipogenic. Autophagy inducer.

Formula: C₁₅H₂₄N₂O₇S
Storage Temperature: Freezer
CAS Number: 133343-34-7

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces lactacystinnaeus strain OM-6519.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in water, methanol, ethanol, DMSO, pyridine, acetonitrile or dimethyl formamide. Insoluble in hexane, benzene, chloroform or ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR