EM574 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: idremcinal, Motilide

AG-CN2-0102-C250 AG-CN2-0102-M001
102987-344EA 152.31 USD
102987-344 102987-346
EM574 ≥95%
EM574

Shows strong gastrointestinal motor stimulating (GMS) activity. Motilin receptor agonist. Stimulates smooth muscle cell contraction. Stimulates cholinergic neurons in a non-vagal pathway. Enhances gastrointestinal contractile activity and accelerates gastric emptying. Shows orexigenic activity with affinity for the growth-hormone secretagogue receptor (GHS-R, ghrelin receptor).

Formula: C₃₉H₆₉NO₁₂
Storage Temperature: Freezer
CAS Number: 110480-13-2

Order Now

Specification Test Results

Source/Host Semisynthetic from erythromycin A.
Purity 95% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in methanol, ethanol, DMSO, chloroform, acetone, ethyl acetate or acetonitrile. Insoluble in water or hexane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR