4-α-Phorbol-12-myristate-13-acetate

Supplier: ADIPOGEN CORP MS

AG-CN2-0082-M001
102987-280EA 96.92 USD
102987-280
4-α-Phorbol-12-myristate-13-acetate
4-alpha-Phorbol-12-myristate-13-acetate

Negative control for phorbol ester activation of protein kinase C and studies with PMA (AG-CN2-0010). Might be a reasonable PKC-independent TRPV4 channels agonist.

Formula: C₃₆H₅₆O₈
MDL Number: MFCD00153860
CAS Number: 63597-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR