Cyclosporin A

Supplier: ADIPOGEN CORP MS

AG-CN2-0079-M100 AG-CN2-0079-5100 AG-CN2-0079-G001
102987-270EA 69.23 USD
102987-270 102987-272 102987-274
Cyclosporin A
Proteins and Peptides

Potent immunosuppressant (same as FK-506 and rapamycin)


Source/Host: Isolated from Tolypocladium inflatum.
Purity: >98%
Appearance/Color: White solid.
Solubility: Soluble in DMSO, ethanol, methanol, dichlormethane, chloroform or acetone. Insoluble in water.


Forms a complex with cyclophilin. Inhibits the activity of the calcium/calmodulin-dependent protein phosphatase 2B (PP2B; calcineurin). Prevents the dephosphorylation of nuclear factor of activated T cells (NFAT) transcription factor, leading to disruption of T cell activation. Suppresses proliferation of cytotoxic T cells and inhibits the production of T cell-derived mediators such as interleukin-2 (IL-2). Prevents rejection of transplanted organs. Anti-inflammatory compound in the treatment of several inflammatory skin diseases (e.g. atopic dermatitis) and with potential anti-rheumatic activity (rheumatoid arthritis). Antibacterial. Antifungal. Antiparasitic. Apoptosis inhibitor. Inhibits the mitochondrial permeability transition pore (MPTP) from opening, thus inhibiting cytochrome c release. NF-kappaB suppressor by induction of unfolded protein response (UPR). Anti-cancer compound. Apoptosis and autophagy inducer. Inhibits nitric oxide synthesis induced by interleukin-1alpha, lipopolysaccharides and TNF-alpha. Potently induces highly cardiogenic progenitors from embryonic stem (ES) cells.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR