Fostriecin sodium salt ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (6R)-5,6-Dihydro-6-[(1E,3R,4R,6R,7Z,9Z,11E)-3,6,13-trihydroxy-3-methyl-4-(phosphonooxy)-1,7,9,11-tridecatetraenyl]-2H-pyran-2-one sodium salt, Fostriecin

AG-CN2-0057-C010
102987-176EA 228.46 USD
102987-176
Fostriecin sodium salt ≥98%
Fostriecin sodium salt

Antibiotic


Source/Host: Isolated from Streptomyces pulveraceous subsp. fostreus.
Purity: >98% (HPLC)
Appearance/Color: Colorless solid.
Solubility: Soluble in water, methanol or ethanol.


Anticancer compound. Antifungal. Catalytic inhibitor of topoisomerase II (IC50 = 40 µM). Potent protein phosphatase 2A (PP2A) (IC50 = 1.5 nM) and 4 (PP4) (IC50 = 3 nM) inhibitor. Weak protein phosphatase type 1 (PP1) inhibitor (IC50=131µM). No apparent inhibition on PP2B. Mitotic entry checkpoint inhibitor. Mediates cell cycle arrest at G2-M-phase. Ischemia protective. The PP2A binding site is different from that of okadaic acid

Formula: C₁₉H₂₆O₉PNa
Storage Temperature: Freezer
CAS Number: 87860-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR