Mouse Recombinant Fc (Mouse) IgG2a Control (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
CHI-MF-120IG2A-C100
102981-696EA 456.92 USD
102981-696
Mouse Recombinant Fc (Mouse) IgG2a Control (from HEK293 Cells)
Proteins and Peptides
This product should be used as the control protein for activity tests of fusion proteins including mouse target proteins and mouse IgG2a-Fc domains.