Mouse Recombinant IL-6R (from CHO Cells)
Supplier: ADIPOGEN CORP MS
CHI-MF-11006R-C050
102981-604EA 456.92 USD
102981-604
Mouse Recombinant IL-6R (from CHO Cells)
Proteins and Peptides
Interleukin 6 receptor (IL-6R; CD126) is a type I cytokine receptor, which is a protein complex consisting of an IL-6 receptor subunit (IL-6R) and interleukin 6 signal transducer glycoprotein 130 (gp130). It is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response. Dysregulated production of IL-6 and IL-6R are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.