Human Recombinant CD160 (from CHO cells)
Supplier: ADIPOGEN CORP MS
CHI-HF-210CD160-C100
102981-496EA 498.46 USD
102981-496
Human Recombinant CD160 (from CHO cells)
Proteins and Peptides
CD160 is a GPI-anchored lymphocyte surface receptor in which expression is mostly restricted to the highly cytotoxic CD56(dim) CD16(+) peripheral blood NK subset. CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 acts as a co-activator receptor for CD3-induced proliferation of CD4+ CD160+ T cells isolated from inflammatory skin lesions. CD160-Fc fusion protein targets a novel costimulatory pathway and prolongs allograft survival.