Mouse Recombinant CCL22 (from CHO cells)
Supplier: ADIPOGEN CORP MS
CHI-MF-110CCL22-C010 CHI-MF-110CCL22-C050
102981-448EA 214.62 USD
102981-448 102981-450
Mouse Recombinant CCL22 (from CHO cells)
Proteins and Peptides
CCL22 (C-C Motif Chemokine 22; Stimulated T cell chemotactic protein; STCP1) is a CC chemokine initially isolated from clones of monocyte-derived macrophages and binds to the receptor CCR4. Human CCL22 is highly expressed in macrophage and in monocyte-derived dendritic cells and thymus. It is also found in lymph node, appendix, activated monocytes, resting and activated macrophages. May play a role in the trafficking of activated/effector T lymphocytes to inflammatory sites and other aspects of activated T lymphocyte physiology. CCL22 has been shown to induce chemotaxis or Ca2+ mobilization in dendritic cells, IL-2 activated NK cells and activated T lymphocytes.