7β-Hydroxycholesteryl bishemisuccinate diethanolamine salt ≥99%

Supplier: ADIPOGEN CORP MS
Warning

CDX-H0123-M010 CDX-H0123-M050 CDX-H0123-M250
102981-236EA 271.38 USD
102981-236 102981-238 102981-240
7β-Hydroxycholesteryl bishemisuccinate diethanolamine salt ≥99%
7beta-Hydroxycholesteryl bishemisuccinate diethanolamine salt

Water-soluble derivatives of 7beta-hydroxycholesterol. Anticancer compound, inducing apoptosis in various solid tumor cell lines.

Formula: C₃₅H₅₄O₈·2C₂H₇NO
Storage Temperature: Freezer
CAS Number: 105449-93-2

Order Now

Specification Test Results

Source/Host Synthetic
Purity 99% (HPLC)
Appearance/Color White powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR