Palmarumycin C3

Supplier: ADIPOGEN CORP MS

BVT-0066-M001
102981-194EA 180 USD
102981-194
Palmarumycin C3
Palmarumycin C3

Palmarumycin C3-5,8-quinone is an oxidation product of palmarumycin C3.


  • Spirobisnaphthalene derivative
  • Oxidation product


Spirobisnaphthalene derivative. Oxidation product of palmarumycin C3. This compound has not yet been thoroughly tested for its biological activity.

Formula: C₂₀H₁₂O₆
MW: 348.3 g/mol
CAS Number: 159934-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR