NAPRTase (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40T-0104-C050
102980-444EA 540 USD
102980-444
NAPRTase (from E. coli)
Enzymes
Nicotinate Phosphoribosyltransferase (NAPRTase) is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN)
  • UniProt link O32090: NAPRTase (B. subtilis)
It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.