Mouse Recombinant PARP-2 (from Insect Cells (Sf21))
Supplier: ADIPOGEN CORP MS
AG-40T-0012-C010
102980-362EA 360 USD
102980-362
Mouse Recombinant PARP-2 (from Insect Cells (Sf21))
Proteins and Peptides
PARP-2 is involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks.