Human Recombinant FGF-2 (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40B-0150-C010 AG-40B-0150-3010
102980-318EA 138.46 USD
102980-318 102980-320
Human Recombinant FGF-2 (from E. coli)
Proteins and Peptides
FGF-2 (Basic fibroblast growth factor) belongs to the FGF superfamily that is made up of 22 different fibroblast growth factor genes. FGF-2 and FGF-1 (acidic FGF) are unique in that they do not follow the conventional signal sequence for secretion. FGF-2 is found in five isoforms via alternative initiation of translation. Signaling of FGF-2 occurs through the high-affinity tyrosine kinase receptors FGFR1-4. FGF-2 is a potent wide-spectrum mitogen whose overexpression is associated with immortalization and unregulated cell proliferation in many tumors. FGF-2 functions in angiogenesis, cell survival, tissue repair, embryonic development and inflammation. FGF-2 is widely used to maintain the self-renewal of human embryonic stem cells (ESCs) and to induce pluripotent stem cells. FGF-2 is an established neurogenic factor for proliferation and differentiation of multipotent neural stem cells both during development and in the adult mouse brain as well.