Human Recombinant TACI (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0079-C050 AG-40B-0079-3050
102980-132EA 387.69 USD
102980-132 102980-134
Human Recombinant TACI (from HEK293 Cells)
Proteins and Peptides
B-cell activating factor belonging to the TNF family receptor (BAFF-R), B-cell maturation antigen (BCMA), and transmembrane activator and calcium modulator and cyclophilin ligand interactor (TACI) play critical functions in promoting B-cell survival by engaging the ligands a proliferation-inducing ligand (APRIL) and/or BAFF. TACI is predominantly expressed on peripheral blood B cells and interacts with the B cell activating factor (BAFF) of ligands and a proliferation inducing ligand (APRIL), which are expressed by numerous cell types, including dendritic cells, macrophages and neutrophils. TNFRSF13B, the gene, which encodes TACI is mutated in nearly 10% of patients with common variable immune deficiency (CVID), an antibody deficiency syndrome characterized by loss of memory B cells and plasma cells.