Mouse BNP-45, AnaSpec
Supplier: Anaspec Inc
BNP-45 represents the 45 amino acids at the C-terminus and the mouse BNP-45 has all the amino acid residues thought essential for NP bioactivity, although sequence identity when studied with other BNP hormones (rat, 64%; dog, 53%; pig, 50%; and human, 44%) was clearly less than the identity among ANF hormones. The threonine 81 residue at which a protein kinase C phosphorylation site is present in the putative mature mouse BNP-45 hormone is not conserved in the rat sequence.
Sequence:SQGSTLRVQQRPQNSKVTHISSCFGHKIDRIGSVSRLGCNALKLL (Disulfide bridge:23 - 39)
MW:4919.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...