3-Isobutyl-1-methylxanthine

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione

AG-CR1-3512-M500 AG-CR1-3512-G001
102516-252EA 135.69 USD
102516-252 102516-254
3-Isobutyl-1-methylxanthine
3-Isobutyl-1-methylxanthine

Cell permeable, competitive, non-specific cAMP and cGMP phosphodiesterase inhibitor. Increases cAMP levels that activate PKA, leading to decreased proliferation, increased differentiation and induction of apoptosis. Enhances differentiation of 3T3-L1 cells. Non-selective adenosine receptor antagonist. Inhibits Ca2+ ion channels. Activates TNF-alpha. Adipogenic. Activates leukotriene synthesis. Reduces inflammation and innate immunity.

Formula: C₁₀H₁₄N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD00005584
CAS Number: 28822-58-4

Order Now

Specification Test Results

Purity 99% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, ethanol or methanol. Insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR