(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one

AG-CR1-0056-M001 AG-CR1-0056-M005 AG-CR1-0056-M025
102516-094EA 41.54 USD
102516-094 102516-096 102516-098
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime

Phosphoinositide-dependent kinase 1 (PDK1) inhibitor. Potent, reversible and ATP-competitive glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) inhibitor. JAK/STAT3 signaling inhibitor. Apoptosis inducer. Potent antiproliferative agent in malignant lymphoid cell. Antimetastatic agent. Potent chemosensitizing agent fighting TRAIL resistant cancer cells.

Formula: C₁₆H₁₀BrN₃O₂
Storage Temperature: Freezer
MDL Number: MFCD08705318
CAS Number: 667463-62-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Dark red solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR