Sirtinol ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(((2-Hydroxynaphthalen-1-yl)methylene)amino)-N-(1-phenylethyl)benzamide

AG-CR1-0055-M001 AG-CR1-0055-M005 AG-CR1-0055-M025
102516-088EA 41.54 USD
102516-088 102516-090 102516-092
Sirtinol ≥98%
Sirtinol

Specific cell permeable SIRT1 (sirtuin 1) inhibitor. Senescence-like growth arrest inducer. Platelet aggregation inhibitor. Apoptosis inducer.

Formula: C₂₆H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00810186
CAS Number: 410536-97-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Off-white to yellow solid.
Solubility Soluble in DMSO, 100% ethanol or dimethyl formamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR