BIX 01294 ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(Hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine, BIX-01294, N-(1-Benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine

AG-CR1-0051-M001 AG-CR1-0051-M005 AG-CR1-0051-M025
102516-072EA 48.46 USD
102516-072 102516-074 102516-076
BIX 01294 ≥98%
BIX 01294

Selective and cell permeable G9a histone-lysine methyltransferase (HMTase) inhibitor. Does not compete with cofactor S-adenosylmethionine. Pluripotent stem cell inducer.

Formula: C₂₈H₃₈N₆O₂
Storage Temperature: Freezer
MDL Number: MFCD14560563
CAS Number: 935693-62-2

Order Now

Specification Test Results

Purity 98% (NMR, HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO (5 mg/ml), 100% ethanol or dimethyl formamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR