bpV(phen) (Potassium hydrate) ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: Potassium bisperoxo(1,10-phenanthroline)oxovanadate (V)

AG-CR1-0042-M005 AG-CR1-0042-M025
102516-044EA 83.08 USD
102516-044 102516-046
bpV(phen) (Potassium hydrate) ≥99%
bpV(phen) (Potassium hydrate)

Potent protein phosphotyrosine phosphatase inhibitor. Insulin receptor kinase (IRK) inducer. Insulin mimetic in vitro and in vivo. Apoptosis inducer. ERK inducer. Mitogen-activated protein kinase phosphatase-1 (MKP-1) inducer. Angiogenesis inhibitor. PTEN inhibitor.

Formula: K[VO(O₂)₂C₁₂H₈N₂]·3H₂O
Storage Temperature: Freezer
CAS Number: 171202-16-7

Order Now

Specification Test Results

Purity >99% (51V-NMR)
Appearance/Color Yellow to orange crystalline solid.
Solubility Soluble in DMSO or water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR