SB-366791 ≥99%
Supplier: ADIPOGEN CORP MS
Synonyms:
4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide
AG-CR1-0034-M005
AG-CR1-0034-M025
102516-012EA
76.15
USD
102516-012
102516-014
SB-366791 ≥99%
SB-366791
Potent and selective TRPV1 antagonist. Glutamatergic synaptic transmission inhibitor.
Formula:
C₁₆H₁₄ClNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD01033818 CAS Number: 472981-92-3 |
Specification Test Results
Purity |
|
Appearance/Color | White to beige crystalline solid. |
Solubility | Soluble in 100% ethanol (warm) or DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...