SB-366791 ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide

AG-CR1-0034-M005 AG-CR1-0034-M025
102516-012EA 76.15 USD
102516-012 102516-014
SB-366791 ≥99%
SB-366791

Potent and selective TRPV1 antagonist. Glutamatergic synaptic transmission inhibitor.

Formula: C₁₆H₁₄ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD01033818
CAS Number: 472981-92-3

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color White to beige crystalline solid.
Solubility Soluble in 100% ethanol (warm) or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR