SIN-1 chloride ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0027-M010 AG-CR1-0027-M050
102515-974EA 41.54 USD
102515-974 102515-976
SIN-1 chloride ≥98% (by NMR)
SIN-1 chloride

Nitric oxide (NO) donor. Generates both superoxide anion and nitric oxide that spontaneously form peroxynitrite. Platelet aggregation inhibitor. Guanylyl cyclase activator. Cytotoxic. Positive ionotropic. Vasodilator.

Formula: C₆H₁₁ClN₄O₂
Storage Temperature: Freezer
CAS Number: 16142-27-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in 100% ethanol, water or DMSO ( 5 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR