Podophyllotoxin ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: (10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.03,7.011,15]hexadeca-1,3(7),8-trien-12-one, Condylox, PPT, (-)-Podophyllotoxin, Podofilox, Condyline, NSC 24818

AG-CN2-0049-M100
102515-854EA 41.54 USD
102515-854
Podophyllotoxin ≥98%
Podophyllotoxin

Potent microtubule assembly inhibitor. Anticancer compound. Cell death inducer. DNA topoisomerase II inhibitor. Cell cycle inhibitor at the metaphase. Antiviral and antihelminthic.

Formula: C₂₂H₂₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00075290
CAS Number: 518-28-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Podophyllum emodi rhizomes.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in chloroform, acetone, ethyl acetate or ethanol. Insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR