Latrunculin B ≥96%

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0031-M001
102515-788EA 276.92 USD
102515-788
Latrunculin B ≥96%
Latrunculin B

Cell permeable marine toxin. Disrupts microfilament-mediated processes. Actin polymerization inhibitor in vitro and in vivo by the formation of a 1:1 complex with monomeric G-actin. Depolymerizes actin filaments (F-actin). Less potent than Latrunculin A. May have fewer unwanted effects than latrunculin A and may be preferred for short-term studies. Acts via a different mechanism than cytochalasins.

Formula: C₂₀H₂₉NO₅S
Storage Temperature: Freezer
CAS Number: 76343-94-7

Order Now

Specification Test Results

Source/Host Isolated from Latrunculia magnifica.
Purity 96% (HPLC)
Appearance/Color White to light yellow solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR