Bilobalide ≥95% (by 1H-NMR)

Supplier: ADIPOGEN CORP MS

AG-CN2-0026-M010 AG-CN2-0026-M050
102515-764EA 90 USD
102515-764 102515-766
Bilobalide ≥95% (by 1H-NMR)
Bilobalide

Neuroprotective. Mitochondrial gene expression regulator. ROS scavenger. Competitive GABA(A) receptor antagonist. Apoptosis inhibitor. CREB phosphorylation enhancer. Stimulates neurogenesis and synaptogenesis. Hepatic cytochrome P450 inducer. Activates the phosphatidylinositol 3-kinase (PI3K) dependent pathway. Potent anti-inflammatory and antihyperalgesic agent.

Formula: C₁₅H₁₈O₈
Storage Temperature: Freezer
MDL Number: MFCD00238547
CAS Number: 33570-04-6

Order Now

Specification Test Results

Source/Host Isolated from Ginko biloba.
Purity 95% (1H-NMR)
Appearance/Color White solid.
Solubility Soluble in ethanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR