X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Supplier: ADIPOGEN CORP MS

AG-CC1-0003-M050 AG-CC1-0003-M250 AG-CC1-0003-G001 AG-CC1-0003-G005
102515-648EA 34.62 USD
102515-648 102515-650 102515-652 102515-654
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Molecular biology reagent. Chromogenic substrate for beta-galactosidase. Used in gene cloning together with IPTG (Prod. No. AG-CC1-0002) to indicate whether a cell expresses the beta-galactosidase enzyme, which is encoded by the lacZ gene, in a technique called blue/white screening. Distinguishes between recombinant and non-recombinant plasmids carrying the beta-galactosidase gene. Used in two-hybrid analysis. X-Gal is used to determine E. coli and coliform content in drinking water samples.

Formula: C₁₄H₁₅BrClNO₆
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now

Specification Test Results

Purity 99% (HPLC)
Appearance/Color White to off-white powder. Clear, colourless to light yellow in solution.
Solubility Soluble in dimethyl formamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR