Human Recombinant Contactin-6 H:Fc H R (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0156-C010 AG-40A-0156-C050
102515-252EA 249.23 USD
102515-252 102515-254
Human Recombinant Contactin-6 H:Fc H R (from HEK293 Cells)
Proteins and Peptides
Contactin-6 is a member of the contactins, which mediate cell surface interactions during nervous system development. Contactin-6 has a key role in the formation of glutamatergic, but not GABAergic, synapses during postnatal development of the hippocampal formation as well as the cerebellum. It is involved in oligodendrocytes generation by acting as a ligand of Notch1. It is also involved in motor coordination.