Human Recombinant ANGPTL7 (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0060-C010 AG-40A-0060-C050
102514-886EA 304.62 USD
102514-886 102514-888
Human Recombinant ANGPTL7 (from HEK293 cells)
Proteins and Peptides
ANGPTL7 (Angiopoietin-like protein 7) is expressed in various tissues and regulates angiogenesis. Human ANGPTL7 was characterized as a potent target gene of the WNT/beta-catenin signaling pathway, and is a pharmacogenomics target in the fields of oncology and regenerative medicine. Overexpression of ANGPTL7 increases collagen expression and might exert a pathogenic role in glaucoma.