Human Recombinant ST2 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0059-C050
102514-884EA 581.54 USD
102514-884
Human Recombinant ST2 (from HEK293 Cells)
Proteins and Peptides
The ST2 (Interleukin-1 receptor-like 1; Interleukin-33 receptor) gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form (ST2) and a longer, transmembrane form (ST2L) by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signaling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.