Rat Recombinant RBP4 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0049-C010 AG-40A-0049-C050
102514-850EA 207.69 USD
102514-850 102514-852
Rat Recombinant RBP4 (from HEK293 Cells)
Proteins and Peptides
Retinol binding protein 4 (RBP4; RBP) is a 21kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol (vitamin A) to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 (GLUT4) suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.