Ascolactone ≥98%

Supplier: ADIPOGEN CORP MS
Danger

BVT-0340-C500 BVT-0340-M001
102513-778EA 131.54 USD
102513-778 102513-780
Ascolactone ≥98%
Ascolactone

Antifungal. Phytotoxic.

Formula: C₁₆H₃₀O₄
Storage Temperature: Freezer
CAS Number: 757995-43-0
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Ascochyta sp.
Purity 98% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in methylene chloride or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR