Simocyclinone D8 (from Streptomyces antibioticus Tü 6040) ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0290-M001
102513-728EA 124.62 USD
102513-728
Simocyclinone D8 (from Streptomyces antibioticus Tü 6040) ≥97% (by HPLC)
Simocyclinone D8

Simocyclinone D8 is a bacterial DNA gyrase inhibitor.


  • Anti-bacterial (Gram-positive), Anti-tumor compound.
  • Antobiotic
  • DNA gyrase inhibitor


Bacterial DNA gyrase inhibitor: Staphylococcus aureus (IC50= 1.45µM), E.coli (IC50= 0.41µM). Human topoisomerase II (Topo II) inhibitor (IC50= 100µM).


Yellow powder


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO, methanol, acetone, dichloromethane or ethyl acetate

Storage Temperature: Freezer
CAS Number: 301845-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR