Borrelidin (from Streptomyces species) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0098-C500 BVT-0098-M001 BVT-0098-M005
102513-544EA 110.77 USD
102513-544 102513-546 102988-886
Borrelidin (from Streptomyces species) ≥98% (by HPLC)
Borrelidin

Borrelidin is a bacterial, protozoan and mammalian threonyl-tRNA synthetase (THrRS) inhibitor.


  • Macrolide antibiotic
  • Inhibitor of potato scab disease
  • Cyclin-dependent kinase (CDK) inhibitor


Antiangiogenic (IC50 = 0.8 nM) and anticancer agent. Induces the collapse of formed capillary tubes in a dose-dependent fashion. In HUVECs, the capillary tube collapsing activity is mediated by the activation of caspases-3 and -8 and induction of apoptosis. Blocks the formation of spontaneous lung metastases of B16-BL6 melanoma cells. Antiviral. Potent antimalarial agent (IC50 = 0.97 nM).


White to off-white powder


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or methanol.

Formula: C₂₈H₄₃NO₆
Storage Temperature: Freezer
CAS Number: 7184-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR