Pyrrolcarbonyltaloside (from Kitasatospora sp. Goe M1) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0095-M001
102513-536EA 110.77 USD
102513-536
Pyrrolcarbonyltaloside (from Kitasatospora sp. Goe M1) ≥98% (by HPLC)
Pyrrolcarbonyltaloside

Pyrrolcarbonyltaloside is an antibiotic.


  • Antibiotic
  • Shows weak cytotoxic activity
  • Anti-parasitic activity


Antibiotic. Shows weak cytotoxic activity against different tumor cell lines. Shows antiparasitic activity.


Brown solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or methanol

Formula: C₁₁H₁₅NO₆
Storage Temperature: Freezer
CAS Number: 912539-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR