Bafilomycin C1 (from Streptomyces sp. Gö 14F) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0068-M001
102513-502EA 207.69 USD
102513-502
Bafilomycin C1 (from Streptomyces sp. Gö 14F) ≥98% (by HPLC)
Bafilomycin C1

Bafilomycin C1 is a potential anti-osteoporotic agent in treating bone lytic diseases.


  • Antibiotic
  • Neuroprotection inducer
  • Anti-osteoporotic agent in treating bone lytic diseases


Antibiotic. Specific vacuolar-type H+-ATPase inhibitor. Antibacterial, antifungal, insecticidal and antihelmintic. Potential anti-osteoporotic agent in treating bone lytic diseases. Neuroprotection inducer.


Yellow powder


After reconstitution protect from light at -20°C. Stable for at least 1 year after receipt when stored at -20°C.


Soluble in 100% ethanol, methanol, DMSO or dimethyl formamide.

Formula: C₃₉H₆₀O₁₂
Storage Temperature: Freezer
CAS Number: 88979-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR