Aspinonene ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0035-M001
102513-480EA 110.77 USD
102513-480
Aspinonene ≥98% (by HPLC)
Aspinonene

Multifunctional fungal secondary metabolite. Related to aspyrone (Prod. No. BVT-0029).

Formula: C₉H₁₆O₄
Storage Temperature: Freezer
CAS Number: 157676-96-5

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus ochraceus (DSM-7428).
Purity 98% (HPLC)
Appearance/Color Oil.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR