Human CD86 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-589-020
102513-382EA 617.54 USD
102513-382
Human CD86 (from CHO cells)
Proteins and Peptides
Human CD86 (B7-2) is a costimulating ligand for CD28 and CD152 (CTLA-4). CD86 is expressed on activated B cells and blood monocytes.