Human CD268 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-524-020
102513-304EA 617.54 USD
102513-304
Human CD268 (from CHO cells)
Proteins and Peptides
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.