Human CD269 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-519-030
102513-298EA 839.08 USD
102513-298
Human CD269 (from CHO cells)
Proteins and Peptides
BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. Its downstream signaling is dependent on NF-kappaB and JNK.