Human CD134 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-513-820
102513-278EA 617.54 USD
102513-278
Human CD134 (from CHO cells)
Proteins and Peptides
Human CD134 (OX40; ACT35) is an activation-associated antigen which is predominantly expressed on activated CD4 positive cells. CD134 antigen is a member of the TNF receptor family of molecules and may be involved with regulating T cell-dependent B cell proliferation and differentiation.