Human CD252 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-512-820
102513-272EA 617.54 USD
102513-272
Human CD252 (from CHO cells)
Proteins and Peptides
Human CD252 (CD134L; OX40 ligand) is a type II membrane protein with homology to TNF which is expressed on activated B cells and activated endothelial cells. CD252 binds to CD134 present on activated T cells, providing a costimulatory signal. Blockade of this interaction in mouse abrogated immunological effects in several models of inflammation and rejection. CD252-muCD8 fusion protein binds to CD134 positive cells in flow cytometry.