Human CD137 (from CHO cells)
Supplier: ADIPOGEN CORP MS
ANC-502-820
102513-220EA 617.54 USD
102513-220
Human CD137 (from CHO cells)
Proteins and Peptides
Human CD137 (4-1BB) is expressed on activated T cells within 24-48 hours of activation. CD137 is a type I membrane protein and a member of the TNF receptor superfamily. CD137 appears to be important for T cell proliferation and survival and induces monocyte activation.