Tadalafil

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: (6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione

S1512-50MG
101763-000EA 181.54 USD
101763-000 101763-002 101763-004
Tadalafil
Tadalafil

Tadalafil is a PDE-5 inhibitor with IC50 of 1.8 nM.

Formula: C₂₂H₁₉N₃O₄
MDL Number: MFCD07771966
CAS Number: 171596-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR