Linezolid (synthetic)

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: Zyvox, (S)-N-((3-(3-fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl)acetamide

S1408-100MG
101760-102EA 746.15 USD
101760-102
Linezolid (synthetic)
Linezolid

Linezolid (Zyvox) is a synthetic antibiotic used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics


A member of the oxazolidinone class of drugs, Linezolid is active against most Gram-positive bacteria that cause disease, including streptococci, vancomycin-resistant enterococci (VRE), and methicillin-resistant Staphylococcus aureus (MRSA). Marketed by Pfizer as Zyvox.

Formula: C₁₆H₂₀FN₃O₄
MW: 337.4 g/mol
MDL Number: MFCD00937825
CAS Number: 165800-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR