Cladribine 99.5%

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: (2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol, CdA, 2-Chloro-2'-deoxyadenosine, NSC105014, RWJ 26251

S1199-25MG
101757-882EA 126.15 USD
101757-882
Cladribine 99.5%
Cladribine

Cladribine is an adenosine deaminase inhibitor for U266, RPMI8226, and MM1.S cells with IC50 of approximately 2.43, 0.75 and 0.18 μM, respectively


  • Assay type: Cytotoxicity assay
  • Powder
  • Solubility: 57 mg/mL (DMSO)


Cladribine exerts remarkable activity in hairy cell leukemia (HCL), a chronic B-cell lymphoproliferative disorder, producing prolonged complete remissions. Cladribine induces accumulation of DNA strand breaks, and subsequently activates the tumor suppressor p53 in lymphocytes. Cladribine may modulate STAT3 activity in MM cells. Cladribine inhibits proliferation/survival of U266, RPMI8226 and MM1.S cells in a dose-dependent manner. While U266 is the least sensitive cell line, MM1.S is the most sensitive one to cladribine. Treatment with cladribine gradually increases the percentage of cells in the G1 phase of the cell cycle and reduces the percentage of cells in S phase. Cladribine appears to increase G2-M phase in U266 cells upon 24 hour-treatment. A dose-dependent increase in apoptosis induced by cladribine is seen in both RPMI8226 and MM1.S cells


Treatment with cladribine at 0.2 μM dramatically induces activation of caspase-3, -8, and -9 and PARP cleavage in a time-dependent manner in MM1.S. Cladribine significantly decreases the phospho-STAT3 (P-STAT3) levels in a dose-dependent manner, but has no effect on the total STAT3 protein levels. Cladribine possesses concentration-dependent apoptosis-inducing potential in the HSB2 cells. Cladribine inhibits growth of primary mast cell (MC) and the MC line HMC-1 in a dose-dependent manner, with lower IC50 values recorded in HMC-1.2 cells harboring KIT D816V compared to HMC-1.1 cells lacking KIT D816V. Cladribine decreases the migratory capacity of CD14+ monocytes, as well as of CD4+ and CD8+ T lymphocytes.


Cladribine is primarily active in lymphoid tissues.

Formula: C₁₀H₁₂ClN₅O₃
MW: 285.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00153939
CAS Number: 4291-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR