Macrophage Detachment Solution DXF, PromoCell
Supplier: Promocell, Inc.
C-41330
10175-308EA 187.13 USD
10175-308
Macrophage Detachment Solution DXF, PromoCell
Cell Separation Media
The Macrophage Detachment Solution DXF is a non-enzymatic reagent especially optimized for the gentle detachment of strongly adherent macrophages.
  • Animal component-free
Macrophages are strongly adherent and are not effectively dislodged from culture surfaces by standard enzymatic cell dissociation reagents. Indeed, these cells are highly sensitive to improper dissociation procedures. It was especially designed for the gentle release of adherent macrophages and guarantees the best possible cell viability, even after prolonged exposure times. Unlike enzyme-based solutions the PromoCell Macrophage Detachment Solution DXF does not alter cell surface proteins and neutralization is not necessary.